MAT154602889 Matrix Scientific

3-(5,6,7,8-Tetrahydro-1-naphthalenyloxy)piperidine hydrochloride

  • MDL number MFCD09879206
  • Cas Number : 1185301-43-2
  • Molecular Weight : 267.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023167.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023167.html
Availability details for
Quantity
  • Availability
  • Add to Cart