MAT154602909 Matrix Scientific

Methyl (2S,4S)-4-(3,4-dichlorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

  • MDL number MFCD09879255
  • Cas Number : 1217735-53-9
  • Molecular Weight : 326.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023646.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023646.html
Availability details for
Quantity
  • Availability
  • Add to Cart