MAT158502006 Matrix Scientific

5-Chloro-2-(pyridin-3-yloxy)-phenylaminedihydrochloride

  • MDL number MFCD09997046
  • Cas Number : 1185121-16-7
  • Molecular Weight : 293.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019900.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019900.html
Availability details for
Quantity
  • Availability
  • Add to Cart