MAT158502007 Matrix Scientific

3-Chloro-2-propoxy-phenylamine hydrochloride

  • MDL number MFCD09997047
  • Cas Number : 1185057-51-5
  • Molecular Weight : 222.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020217.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020217.html
Availability details for
Quantity
  • Availability
  • Add to Cart