MAT158502012 Matrix Scientific

3-[(4-Phenoxyphenoxy)methyl]piperidinehydrochloride

  • MDL number MFCD09997058
  • Cas Number : 1185296-62-1
  • Molecular Weight : 319.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023016.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023016.html
Availability details for
Quantity
  • Availability
  • Add to Cart