MAT161065961 Matrix Scientific

3-Thiophen-2-yl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02677688
  • Cas Number : 26033-27-2
  • Molecular Weight : 178.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-71
Country of Origin USA
Product Link http://www.matrixscientific.com/008342.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008342.html
Availability details for
Quantity
  • Availability
  • Add to Cart