MAT161067831 Matrix Scientific

Diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate

  • MDL number MFCD00005450
  • Cas Number : 4815-30-9
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108
Storage temp. Ambient
Country of Origin US
Product Link http://www.matrixscientific.com/028819.html

Safety Information

MSDS http://www.matrixscientific.com/028819.html
Availability details for
Quantity
  • Availability
  • Add to Cart