MAT161068582 Matrix Scientific

2-Chloro-N-1-naphthylpropanamide

  • MDL number MFCD06654954
  • Cas Number : 22302-58-5
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029570.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029570.html
Availability details for
Quantity
  • Availability
  • Add to Cart