MAT161068640 Matrix Scientific

2-[(Chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxamide

  • MDL number MFCD01355935
  • Cas Number : 551903-76-5
  • Molecular Weight : 246.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029628.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029628.html
Availability details for
Quantity
  • Availability
  • Add to Cart