MAT161068656 Matrix Scientific

Methyl 3-[(chloroacetyl)amino]thiophene-2-carboxylate

  • MDL number MFCD00120696
  • Cas Number : 146381-88-6
  • Molecular Weight : 233.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029644.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029644.html
Availability details for
Quantity
  • Availability
  • Add to Cart