MAT161068774 Matrix Scientific

2-(3,4-Dichlorophenoxy)propanohydrazide

  • MDL number MFCD00210182
  • Cas Number : 588673-62-5
  • Molecular Weight : 249.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029762.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029762.html
Availability details for
Quantity
  • Availability
  • Add to Cart