MAT161068936 Matrix Scientific

2,5-Dimethoxybenzenesulfonohydrazide

  • MDL number MFCD03423368
  • Cas Number : 19116-91-7
  • Molecular Weight : 232.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029924.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029924.html
Availability details for
Quantity
  • Availability
  • Add to Cart