MAT161068937 Matrix Scientific

3,4-Dimethoxybenzenesulfonohydrazide

  • MDL number MFCD03423369
  • Cas Number : 23095-32-1
  • Molecular Weight : 232.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029925.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029925.html
Availability details for
Quantity
  • Availability
  • Add to Cart