MAT161069000 Matrix Scientific

N-(3-Acetylphenyl)-5-methylisoxazole-3-carboxamide

  • MDL number MFCD03419834
  • Cas Number : 784086-73-3
  • Molecular Weight : 244.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029988.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029988.html
Availability details for
Quantity
  • Availability
  • Add to Cart