MAT161069003 Matrix Scientific

N-(4-Acetylphenyl)-5-methylthiophene-3-carboxamide

  • MDL number MFCD03419970
  • Cas Number : 895920-84-0
  • Molecular Weight : 259.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/029991.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/029991.html
Availability details for
Quantity
  • Availability
  • Add to Cart