MAT161069012 Matrix Scientific

N-(3-Acetylphenyl)-5-ethylthiophene-3-carboxamide

  • MDL number MFCD03419979
  • Cas Number : 895935-82-7
  • Molecular Weight : 273.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030000.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030000.html
Availability details for
Quantity
  • Availability
  • Add to Cart