MAT161069045 Matrix Scientific

1-(4-Propylphenyl)propan-1-one

  • MDL number MFCD01993684
  • Cas Number : 56147-30-9
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/030033.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030033.html
Availability details for
Quantity
  • Availability
  • Add to Cart