MAT161069128 Matrix Scientific

N-(3-Acetylphenyl)cyclobutanecarboxamide

  • MDL number MFCD02176923
  • Cas Number : 109920-59-4
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030116.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030116.html
Availability details for
Quantity
  • Availability
  • Add to Cart