MAT161069146 Matrix Scientific

2-(2-Acetylphenoxy)acetamide

  • MDL number MFCD02245031
  • Cas Number : 445232-49-5
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030134.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030134.html
Availability details for
Quantity
  • Availability
  • Add to Cart