MAT161069173 Matrix Scientific

2-Fluoro-4-isocyanato-1-methoxybenzene

  • MDL number MFCD09971932
  • Cas Number : 221218-33-3
  • Molecular Weight : 167.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/030161.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030161.html
Availability details for
Quantity
  • Availability
  • Add to Cart