MAT161069185 Matrix Scientific

2-Chloro-1-fluoro-4-isocyanatobenzene

  • MDL number MFCD00037037
  • Cas Number : 50529-33-4
  • Molecular Weight : 171.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030173.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030173.html
Availability details for
Quantity
  • Availability
  • Add to Cart