MAT161069189 Matrix Scientific

1-Isocyanato-2-methoxy-4-nitrobenzene

  • MDL number MFCD00037066
  • Cas Number : 33484-67-2
  • Molecular Weight : 194.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030177.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030177.html
Availability details for
Quantity
  • Availability
  • Add to Cart