MAT161069201 Matrix Scientific

Methyl 2-isocyanatothiophene-3-carboxylate

  • MDL number MFCD09971935
  • Cas Number : 50502-27-7
  • Molecular Weight : 183.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030189.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030189.html
Availability details for
Quantity
  • Availability
  • Add to Cart