MAT161069294 Matrix Scientific

5-Ethyl-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03423403
  • Cas Number : 869944-96-7
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030282.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030282.html
Availability details for
Quantity
  • Availability
  • Add to Cart