MAT180692530 Matrix Scientific

5-Bromo-2-(2-methoxyethoxy)aniline hydrochloride

  • MDL number MFCD09997940
  • Cas Number : 1185300-88-2
  • Molecular Weight : 282.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022883.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022883.html
Availability details for
Quantity
  • Availability
  • Add to Cart