MAT180692606 Matrix Scientific

5-(3,4-Dichlorophenyl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01940420
  • Cas Number : 725217-53-8
  • Molecular Weight : 260.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030385.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030385.html
Availability details for
Quantity
  • Availability
  • Add to Cart