MAT180692607 Matrix Scientific

5-(3,4-Dichlorophenyl)-4-ethyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03033548
  • Cas Number : 660417-18-5
  • Molecular Weight : 274.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030386.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030386.html
Availability details for
Quantity
  • Availability
  • Add to Cart