MAT180692708 Matrix Scientific

4-Allyl-5-pyridin-2-yl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03945742
  • Cas Number : 91813-63-7
  • Molecular Weight : 218.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030487.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030487.html
Availability details for
Quantity
  • Availability
  • Add to Cart