MAT180692859 Matrix Scientific

4-Methyl-5-(5-methyl-4-phenylthien-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04968924
  • Cas Number : 847503-30-4
  • Molecular Weight : 287.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030638.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030638.html
Availability details for
Quantity
  • Availability
  • Add to Cart