MAT180692891 Matrix Scientific

7-Amino-2-methyl-3-phenylpyrazolo-[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD09971986
  • Cas Number : 956576-87-7
  • Molecular Weight : 268.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030670.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030670.html
Availability details for
Quantity
  • Availability
  • Add to Cart