MAT180692921 Matrix Scientific

2,2'-(2,2'-(Butane-1,4-diyl)bis(1H-benzo[d]-imidazole-2,1-diyl))diacetic acid

  • MDL number MFCD03270836
  • Cas Number : 445416-61-5
  • Molecular Weight : 406.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030700.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030700.html
Availability details for
Quantity
  • Availability
  • Add to Cart