MAT180692952 Matrix Scientific

[2-(Methylsulfonyl)-1H-benzimidazol-1-yl]-acetic acid

  • MDL number MFCD06755483
  • Cas Number : 891450-28-5
  • Molecular Weight : 254.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030731.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030731.html
Availability details for
Quantity
  • Availability
  • Add to Cart