MAT180693007 Matrix Scientific

2-(2-Phenylethoxy)benzaldehyde

  • MDL number MFCD03820021
  • Cas Number : 93898-91-0
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030786.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030786.html
Availability details for
Quantity
  • Availability
  • Add to Cart