MAT180693030 Matrix Scientific

3-(Isobutyrylamino)benzoic acid

  • MDL number MFCD00227316
  • Cas Number : 28533-44-0
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030809.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030809.html
Availability details for
Quantity
  • Availability
  • Add to Cart