MAT180693036 Matrix Scientific

3-Chloro-N-(thien-2-ylmethyl)propanamide

  • MDL number MFCD03394727
  • Cas Number : 21403-26-9
  • Molecular Weight : 203.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030815.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030815.html
Availability details for
Quantity
  • Availability
  • Add to Cart