MAT180693059 Matrix Scientific

4-(3,4-Dimethoxyphenethyl)-5-(1-(ethylsulfonyl)-piperidin-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08569921
  • Cas Number : 932807-05-1
  • Molecular Weight : 440.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/030838.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030838.html
Availability details for
Quantity
  • Availability
  • Add to Cart