MAT180693087 Matrix Scientific

1-Benzyl-2,3-dimethyl-1H-indole-5-carboxylic acid

  • MDL number MFCD00419414
  • Cas Number : 313498-12-3
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/030866.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030866.html
Availability details for
Quantity
  • Availability
  • Add to Cart