MAT180693110 Matrix Scientific

6-(Difluoromethyl)-4-methyl-1H-pyrazolo[3,4-b]-pyridin-3-amine

  • MDL number MFCD09431203
  • Cas Number : 925146-05-0
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/030889.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030889.html
Availability details for
Quantity
  • Availability
  • Add to Cart