MAT180693134 Matrix Scientific

2-(4-Bromo-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD04969759
  • Cas Number : 51292-42-3
  • Molecular Weight : 219.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030913.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030913.html
Availability details for
Quantity
  • Availability
  • Add to Cart