MAT180693173 Matrix Scientific

2-Hydrazino-4-methyl-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD05861708
  • Cas Number : 123024-52-2
  • Molecular Weight : 192.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030952.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030952.html
Availability details for
Quantity
  • Availability
  • Add to Cart