MAT180693194 Matrix Scientific

4-Bromo-1-ethyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD02090837
  • Cas Number : 514800-97-6
  • Molecular Weight : 219.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030973.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030973.html
Availability details for
Quantity
  • Availability
  • Add to Cart