MAT180693225 Matrix Scientific

3-(2-Aminoethyl)-2-phenylquinazolin-4(3H)-one

  • MDL number MFCD09037244
  • Cas Number : 62838-20-4
  • Molecular Weight : 265.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/031004.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031004.html
Availability details for
Quantity
  • Availability
  • Add to Cart