MAT180693298 Matrix Scientific

3-(Aminosulfonyl)thiophene-2-carboxylic acid

  • MDL number MFCD01760861
  • Cas Number : 59337-97-2
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031077.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031077.html
Availability details for
Quantity
  • Availability
  • Add to Cart