MAT180693497 Matrix Scientific

3-Chloro-2-methoxyaniline hydrochloride

  • MDL number MFCD09763716
  • Cas Number : 1437383-39-5
  • Molecular Weight : 194.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021882.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021882.html
Availability details for
Quantity
  • Availability
  • Add to Cart