MAT180693510 Matrix Scientific

Pyridine-3-sulfonyl chloride hydrochloride

  • MDL number MFCD04035552
  • Cas Number : 42899-76-3
  • Molecular Weight : 214.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-5
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/019750.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/019750.html
Availability details for
Quantity
  • Availability
  • Add to Cart