MAT180694353 Matrix Scientific

N-Neopentyl-3-(trifluoromethyl)aniline

  • MDL number MFCD07787000
  • Cas Number : 887590-46-7
  • Molecular Weight : 231.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031376.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031376.html
Availability details for
Quantity
  • Availability
  • Add to Cart