MAT180694487 Matrix Scientific

2,4-Dichloro-N-(4-methoxyphenethyl)aniline

  • MDL number MFCD10687649
  • Cas Number : 1040689-39-1
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031579.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031579.html
Availability details for
Quantity
  • Availability
  • Add to Cart