MAT180694613 Matrix Scientific

N-(2-Methoxyethyl)-2,2-dimethyl-1-propanamine

  • MDL number MFCD10687812
  • Cas Number : 1040686-84-7
  • Molecular Weight : 145.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/031781.html

Safety Information

hcodes Irritant/Stored under Argon
MSDS http://www.matrixscientific.com/031781.html
Availability details for
Quantity
  • Availability
  • Add to Cart