MAT180694702 Matrix Scientific

N-Ethyl-N-(4-propoxyphenyl)amine

  • MDL number MFCD10687915
  • Cas Number : 59731-93-0
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 145-147� Torr
Product Link http://www.matrixscientific.com/031899.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031899.html
Availability details for
Quantity
  • Availability
  • Add to Cart