MAT180694998 Matrix Scientific

4-(2-Cyclohexylethoxy)-N-neopentylaniline

  • MDL number MFCD10688224
  • Cas Number : 1040693-52-4
  • Molecular Weight : 289.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032224.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032224.html
Availability details for
Quantity
  • Availability
  • Add to Cart